N-(tert-Butoxycarbonyl)-L-serinemethylester ≥95.0%

Supplier: TCI America

Synonyms: Boc-Ser-OMe, N-Boc-L-serine Methyl Ester, BOC-L-Serine methyl ester

B2073-5G B2073-25G
TCB2073-005GEA 32.17 CAD
TCB2073-005G TCB2073-025G
N-(tert-Butoxycarbonyl)-L-serinemethylester ≥95.0%
N-(tert-Butoxycarbonyl)-L-serine methyl ester

CAS Number: 2766-43-0
MDL Number: MFCD00191869
Molecular Formula: C9H17NO5
Molecular Weight: 219.24
Purity/Analysis Method: >95.0% (HPLC,N)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.14
Specific rotation [a]20/D: -18 deg (C=5, MeOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-L-serine Methyl Ester, Boc-Ser-OMe

Formula: C₉H₁₇NO₅
MW: 219.24 g/mol
Density: 1.14 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00191869
CAS Number: 2766-43-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=819e5d9c0d3ec7fac523ef9fe36105052bed2f8c&vid=819e5d9c0d3ec7fac523ef9fe36105052bed2f8c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,N)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR