3-(Benzyloxy)phenol ≥95.0%

Supplier: TCI America

B2070-25G B2070-5G
TCB2070-25GEA 930.62 CAD
TCB2070-25G TCB2070-5G
3-(Benzyloxy)phenol ≥95.0%
3-(Benzyloxy)phenol

CAS Number: 3769-41-3
MDL Number: MFCD00134682
Molecular Formula: C13H12O2
Molecular Weight: 200.24
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 200
Melting point (°C): 50


Other Product Names / Synonyms: Resorcinol Monobenzyl Ether

Formula: C₁₃H₁₂O₂
Boiling Pt: 200 °C
Melting Pt: 50 °C
MDL Number: MFCD00134682
CAS Number: 3769-41-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba1930815c9aa496106eade3b02b776f11358163&vid=ba1930815c9aa496106eade3b02b776f11358163&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR