4-Bromo-2,6-xylenol ≥98.0% for synthesis
Supplier: TCI America
Synonyms:
4-Bromo-2,6-dimethylphenol, 5-Bromo-2-hydroxy-3-xylene
B2060-250G
B2060-25G
TCB2060-250GEA
2008.18
CAD
TCB2060-250G
TCB2060-025G
4-Bromo-2,6-xylenol ≥98.0% for synthesis
4-Bromo-2,6-xylenol
CAS Number: 2374-05-2
MDL Number: MFCD00002314
Molecular Formula: C8H9BrO
Molecular Weight: 201.06
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 79
Other Product Names / Synonyms: 4-Bromo-2,6-xylenol
Formula:
C₈H₉BrO Boiling Pt: 259 °C (1013 hPa) Melting Pt: 79 °C |
MDL Number:
MFCD00002314 CAS Number: 2374-05-2 EINECS: 219-153-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...