5''-Bromo-2,2':5',2''-terthiophene-5-carboxaldehyde ≥94.0%

Supplier: TCI America

B2058-5G B2058-1G
TCB2058-5GEA 1651.01 CAD
TCB2058-5G TCB2058-001G TCB2058-100MG
5''-Bromo-2,2':5',2''-terthiophene-5-carboxaldehyde ≥94.0%
5''-Bromo-2,2':5',2''-terthiophene-5-carboxaldehyde

CAS Number: 161726-69-8
MDL Number: MFCD02093442
Molecular Formula: C13H7BrOS3
Molecular Weight: 355.28
Purity/Analysis Method: >94.0% (GC)
Form: Crystal
Melting point (°C): 164


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 5-Bromo-5''-formyl-2,2':5',2''-terthiophene

Formula: C₁₃H₇BrOS₃
Melting Pt: 164 °C
MDL Number: MFCD02093442
CAS Number: 161726-69-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2b2e136729e33578f353bc83636162d3f1bbe1a&vid=f2b2e136729e33578f353bc83636162d3f1bbe1a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR