(1S,4S)-2-Benzyl-2,5-diazabicyclo[2.2.1]heptane dihydrobromide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B2054-1G B2054-5G
TCB2054-001GEA 202.24 CAD
TCB2054-001G TCB2054-005G
(1S,4S)-2-Benzyl-2,5-diazabicyclo[2.2.1]heptane dihydrobromide ≥98.0% (by titrimetric analysis)
(1S,4S)-2-Benzyl-2,5-diazabicyclo[2.2.1]heptane dihydrobromide

CAS Number: 116258-17-4
MDL Number: MFCD01321292
Molecular Formula: C12H16N2
Molecular Weight: 350.10
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: 16 deg (C=3, 2mol/L NaOH sol.)

Formula: C₁₂H₁₆N₂
MW: 350.10 g/mol
MDL Number: MFCD01321292
CAS Number: 116258-17-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa609e2a9ce310163d7ce8efb4a77eb73dc3a709&vid=aa609e2a9ce310163d7ce8efb4a77eb73dc3a709&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR