Bis(triphenylphosphine)palladium(II) diacetate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Bis(triphenylphosphine)palladium(II) acetate, Bis(acetato)bis(triphenylphosphine)palladium(II), Diacetatobis(triphenylphosphine)palladium(II)

B2042-1G B2042-5G
TCB2042-001GEA 301.78 CAD
TCB2042-001G TCB2042-5G
Bis(triphenylphosphine)palladium(II) diacetate ≥98.0% (by titrimetric analysis)
Bis(triphenylphosphine)palladium(II) diacetate

CAS Number: 14588-08-0
MDL Number: MFCD00010013
Molecular Formula: C40H36O4P2Pd
Molecular Weight: 749.09
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Storage Temperature: <0°C


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Cross-coupling Reaction using Transition Metal Catalysts
Other Product Names / Synonyms: Bis(acetato)bis(triphenylphosphine)palladium(II)

Formula: C₄₀H₃₆O₄P₂Pd
MW: 749.09 g/mol
MDL Number: MFCD00010013
CAS Number: 14588-08-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91eac84173a0dcd5a9b92c039685c557f19e1c21&vid=91eac84173a0dcd5a9b92c039685c557f19e1c21&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR