Allyl benzoate ≥98.0%

Supplier: TCI America

Synonyms: Benzoic acid Allyl Ester, Benzoic Acid Allyl Ester, Allyl Benzoate

B2041-250G B2041-25G
TCB2041-250GEA 676.24 CAD
TCB2041-250G TCB2041-025G
Allyl benzoate ≥98.0%
Allyl benzoate

CAS Number: 583-04-0
MDL Number: MFCD00026104
Molecular Formula: C10H10O2
Molecular Weight: 162.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 242
Specific Gravity (20/20): 1.05


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Benzoic Acid Allyl Ester

Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Boiling Pt: 242 °C
Density: 1.05 (20 °C)
MDL Number: MFCD00026104
CAS Number: 583-04-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76b50bddf03e612f28a4fcf8199adb9db04952b6&vid=76b50bddf03e612f28a4fcf8199adb9db04952b6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR