2,2-Bis(hydroxymethyl)butyric acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America

B2038-500G B2038-25G
TCB2038-500GEA 304.94 CAD
TCB2038-500G TCB2038-025G
2,2-Bis(hydroxymethyl)butyric acid ≥97.0% (by titrimetric analysis)
2,2-Bis(hydroxymethyl)butyric acid

CAS Number: 10097-02-6
MDL Number: MFCD00190088
Molecular Formula: C6H12O4
Molecular Weight: 148.16
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 112
Flash Point (°C): 220


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 2,2-Dimethylolbutyric Acid

Formula: C₆H₁₂O₄
Melting Pt: 112 °C
Flash Pt: 220 °C
MDL Number: MFCD00190088
CAS Number: 10097-02-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efc5e3d57abf41e008b3ed2651d949e5b9194c5a&vid=efc5e3d57abf41e008b3ed2651d949e5b9194c5a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR