Benzylbis(triphenylphosphine)palladium(II) chloride ≥96.0%

Supplier: TCI America

Synonyms: (SP-4-3)-Benzylbis(triphenylphosphine)palladium(II) chloride, trans-Benzyl(chloro)bis(triphenylphosphine)palladium(II), Benzyl(chloro)bis(triphenylphosphine)palladium(II)

B2029-100MG B2029-1G
TCB2029-100MGEA 55.3 CAD
TCB2029-100MG TCB2029-001G
Benzylbis(triphenylphosphine)palladium(II) chloride ≥96.0%
Benzylbis(triphenylphosphine)palladium(II) chloride

CAS Number: 22784-59-4
MDL Number: MFCD00010395
Molecular Formula: C43H37ClP2Pd
Molecular Weight: 757.59
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Storage Temperature: <0°C


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Cross-coupling Reaction using Transition Metal Catalysts
Other Product Names / Synonyms: Benzyl(chloro)bis(triphenylphosphine)palladium(II)

Formula: C₄₃H₃₇ClP₂Pd
MW: 757.59 g/mol
MDL Number: MFCD00010395
CAS Number: 22784-59-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bcc57c31d3952591ae929ade6762f27226ad46a1&vid=bcc57c31d3952591ae929ade6762f27226ad46a1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR