6-Bromoveratraldehyde ≥98.0%

Supplier: TCI America

B2011-5G B2011-25G
TCB2011-005GEA 42.66 CAD
TCB2011-005G TCB2011-025G
6-Bromoveratraldehyde ≥98.0%
6-Bromoveratraldehyde

CAS Number: 5392-10-9
MDL Number: MFCD00003301
Molecular Formula: C9H9BrO3
Molecular Weight: 245.07
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 148


Other Product Names / Synonyms: 6-Bromoveratraldehyde

Formula: C₉H₉BrO₃
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 148 °C
MDL Number: MFCD00003301
CAS Number: 5392-10-9
EINECS: 226-390-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e7d466b8299b1f914b9ed49fe7e95183891d4f0&vid=b52d7b0e9092768190f0d027d618749253ac0a55&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR