4-Bromophthalimide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B2009-25G B2009-5G
TCB2009-25GEA 334.96 CAD
TCB2009-25G TCB2009-005G
4-Bromophthalimide ≥98.0% (by HPLC, titration analysis)
4-Bromophthalimide

CAS Number: 6941-75-9
MDL Number: MFCD00466049
Molecular Formula: C8H4BrNO2
Molecular Weight: 226.03
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 235

Formula: C₈H₄BrNO₂
Melting Pt: 235 °C
MDL Number: MFCD00466049
CAS Number: 6941-75-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a563390fbdfc247eb2e786dc7ddb27c8e09abb45&vid=a563390fbdfc247eb2e786dc7ddb27c8e09abb45&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR