9,10-Anthraquinone-2-carboxylic acid ≥99.0%

Supplier: TCI America

Synonyms: Anthraquinone-2-carboxylic acid, 9,10-Dioxo-9,10-dihydroanthracene-2-carboxylic acid

A0504-25G A0504-5G
TCA0504-025GEA 570.38 CAD
TCA0504-025G TCA0504-5G
9,10-Anthraquinone-2-carboxylic acid ≥99.0%
9,10-Anthraquinone-2-carboxylic acid

CAS Number: 117-78-2
MDL Number: MFCD00001231
Molecular Formula: C15H8O4
Molecular Weight: 252.23
Purity/Analysis Method: >99.0% (T)
Form: Crystal

Formula: C₁₅H₈O₄
MW: 252.22 g/mol
MDL Number: MFCD00001231
CAS Number: 117-78-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ccdd8dfd339a284c87685ab0305dee0a668fe10f&vid=ccdd8dfd339a284c87685ab0305dee0a668fe10f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR