N-(tert-Butoxycarbonyl)-L-serine-β-lactone 98,0

Supplier: TCI America

Synonyms: (S)-ɑ-tert-Butoxycarbonylamino-β-propiolactone, tert-Butyl (S)-(2-oxooxetan-3-yl)carbamate, N-Boc-L-serineβ-Lactone, (S)-ɑ-Boc-amino-β-propiolactone, (S)-3-(tert-Butoxycarbonylamino)-2-oxetanone

B1995-1G B1995-5G
TCB1995-001GEA 732.17 CAD
TCB1995-001G TCB1995-5G
N-(tert-Butoxycarbonyl)-L-serine-β-lactone 98,0
N-(tert-Butoxycarbonyl)-L-serine-beta-lactone

CAS Number: 98541-64-1
MDL Number: MFCD01318414
Molecular Formula: C8H13NO4
Molecular Weight: 187.20
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Specific rotation [a]20/D: -26 deg (C=1, CH3CN)
Storage Temperature: <0°C


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-alpha-Boc-amino-beta-propiolactone, N-Boc-L-serine beta-Lactone, (S)-3-(tert-Butoxycarbonylamino)-2-oxetanone, (S)-alpha-tert-Butoxycarbonylamino-beta-propiolactone

Formula: C₈H₁₃NO₄
MW: 187.20 g/mol
MDL Number: MFCD01318414
CAS Number: 98541-64-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ffae7f38107e46ba9a6885c77b792b64b918d36f&vid=ffae7f38107e46ba9a6885c77b792b64b918d36f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR