4'-tert-Butylbiphenyl-4-carboxylic acid ≥98.0%

Supplier: TCI America

Synonyms: 4'-(tert-Butyl)-[1,1'-biphenyl]-4-carboxylic acid

B1986-1G B1986-5G
TCB1986-1GEA 167.48 CAD
TCB1986-1G TCB1986-005G
4'-tert-Butylbiphenyl-4-carboxylic acid ≥98.0%
4'-tert-Butylbiphenyl-4-carboxylic acid

CAS Number: 5748-42-5
MDL Number: MFCD01631300
Molecular Formula: C17H18O2
Molecular Weight: 254.33
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4-tert-Butylbiphenyl-4'-carboxylic Acid

Formula: C₁₇H₁₈O₂
MW: 254.3200 g/mol
MDL Number: MFCD01631300
CAS Number: 5748-42-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2c557a2b4067e4dbafad1852066a6e10112c6c0&vid=c2c557a2b4067e4dbafad1852066a6e10112c6c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR