2-Acetamidofluorene ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
A0076-25G
A0076-5G
TCA0076-025GEA
635.16
CAD
TCA0076-025G
TCA0076-5G
2-Acetamidofluorene ≥98.0% (by HPLC, total nitrogen)
2-Acetamidofluorene
CAS Number: 53-96-3
MDL Number: MFCD00001116
Molecular Formula: C15H13NO
Molecular Weight: 223.28
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 196
Product Categories: Bioscience, Reagents for Carcinogenic Models
Other Product Names / Synonyms: N-2-Fluorenylacetamide, 2-AAF
Formula:
C₁₅H₁₃NO Melting Pt: 196 °C |
MDL Number:
MFCD00001116 CAS Number: 53-96-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...