Bis(3-ethyl-5-methyl-4-maleimidophenyl)methane ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: Methylenebis(3-ethyl-5-methyl-4-maleimidobenzene)

B1971-25G B1971-500G B1971-100G
TCB1971-025GEA 129.56 CAD
TCB1971-025G TCB1971-500G TCB1971-100G
Bis(3-ethyl-5-methyl-4-maleimidophenyl)methane ≥98.0% (by total nitrogen basis)
Bis(3-ethyl-5-methyl-4-maleimidophenyl)methane

CAS Number: 105391-33-1
MDL Number: MFCD00467754
Molecular Formula: C27H26N2O4
Molecular Weight: 442.52
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 165


Product Categories: Materials Chemistry, Monomers, Diphenylmethanes (for High-Performance Polymer Research)
Other Product Names / Synonyms: Methylenebis(3-ethyl-5-methyl-4-maleimidobenzene)

Formula: C₂₇H₂₆N₂O₄
MW: 442.52 g/mol
Melting Pt: 165 °C
MDL Number: MFCD00467754
CAS Number: 105391-33-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=112be2b90a2e95a16e1e711aff6669562e74bd51&vid=9f0063961a6e10766d4c8fa060fc433b1b2439dc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR