4,4'-Bis(ɑ,ɑ-dimethylbenzyl)diphenylamine ≥98.0%

Supplier: TCI America

Synonyms: 4,4-Bis(α,α-dimethylbenzyl)diphenylamine

B1970-25G B1970-500G
TCB1970-025GEA 50.56 CAD
TCB1970-025G TCB1970-500G
4,4'-Bis(ɑ,ɑ-dimethylbenzyl)diphenylamine ≥98.0%
4,4'-Bis(alpha,alpha-dimethylbenzyl)diphenylamine

CAS Number: 10081-67-1
MDL Number: MFCD00337918
Molecular Formula: C30H31N
Molecular Weight: 405.59
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 100
Flash Point (°C): 263

Formula: C₃₀H₃₁N
MW: 405.59 g/mol
Melting Pt: 100 °C
Flash Pt: 263 °C
MDL Number: MFCD00337918
CAS Number: 10081-67-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02201472bca5bcd4e2ba06366541e40602522a53&vid=02201472bca5bcd4e2ba06366541e40602522a53&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR