9-(1H-Benzotriazol-1-ylmethyl)-9H-carbazole ≥98.0%

Supplier: TCI America

B1967-5G B1967-1G
TCB1967-005GEA 200.03 CAD
TCB1967-005G TCB1967-001G
9-(1H-Benzotriazol-1-ylmethyl)-9H-carbazole ≥98.0%
9-(1H-Benzotriazol-1-ylmethyl)-9H-carbazole

CAS Number: 124337-34-4
MDL Number: MFCD00216631
Molecular Formula: C19H14N4
Molecular Weight: 298.35
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal

Formula: C₁₉H₁₄N₄
MDL Number: MFCD00216631
CAS Number: 124337-34-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=289b24b23ed2ab88ffcd1312750b0bbf9a63ed16&vid=289b24b23ed2ab88ffcd1312750b0bbf9a63ed16&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR