6-(Bromomethyl)-2-pyridinemethanol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
B1963-5G
B1963-1G
TCB1963-5GEA
1094.31
CAD
TCB1963-5G
TCB1963-001G
6-(Bromomethyl)-2-pyridinemethanol ≥98.0% (by GC, titration analysis)
6-(Bromomethyl)-2-pyridinemethanol
CAS Number: 40054-01-1
MDL Number: MFCD01631297
Molecular Formula: C7H8BrNO
Molecular Weight: 202.05
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 80
Other Product Names / Synonyms: 2-(Bromomethyl)-6-(hydroxymethyl)pyridine
Formula:
C₇H₈BrNO Melting Pt: 80 °C |
MDL Number:
MFCD01631297 CAS Number: 40054-01-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...