2,5-Bis(methoxycarbonyl)-3,4-diphenylcyclopentadienone
Supplier: TCI America
Synonyms:
Dimethyl 3,4-Diphenylcyclopentadienone-2,5-dicarboxylate, 3,4-Diphenylcyclopentadienone-2,5-dicarboxylic Acid Dimethyl Ester
B1962-5G
B1962-1G
TCB1962-5GEA
568.8
CAD
TCB1962-5G
TCB1962-001G
2,5-Bis(methoxycarbonyl)-3,4-diphenylcyclopentadienone
2,5-Bis(methoxycarbonyl)-3,4-diphenylcyclopentadienone
CAS Number: 16691-79-5
MDL Number: MFCD00111095
Molecular Formula: C21H16O5
Molecular Weight: 348.35
Form: Crystal
Color: Yellow
Other Product Names / Synonyms: Dimethyl 3,4-Diphenylcyclopentadienone-2,5-dicarboxylate, 3,4-Diphenylcyclopentadienone-2,5-dicarboxylic Acid Dimethyl Ester
Formula:
C₂₁H₁₆O₅ MW: 348.35 g/mol |
MDL Number:
MFCD00111095 CAS Number: 16691-79-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...