trans-3-Bromocinnamic acid ≥99.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B1961-5G B1961-25G
TCB1961-005GEA 93.22 CAD
TCB1961-005G TCB1961-025G
trans-3-Bromocinnamic acid ≥99.0% (by GC, titration analysis)
trans-3-Bromocinnamic acid

CAS Number: 14473-91-7
MDL Number: MFCD00004382
Molecular Formula: C9H7BrO2
Molecular Weight: 227.06
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 180

Formula: C₉H₇BrO₂
Boiling Pt: 323 °C (1013 hPa)
Melting Pt: 180 °C
MDL Number: MFCD00004382
CAS Number: 14473-91-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11155bbe0f22aceb1a327013e0bec9f1a6920236&vid=11155bbe0f22aceb1a327013e0bec9f1a6920236&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR