1,6-Bis(diphenylphosphino)hexane ≥95.0%

Supplier: TCI America

Synonyms: Hexamethylenebis(diphenylphosphine)

B1959-1G B1959-5G
TCB1959-001GEA 56.88 CAD
TCB1959-001G TCB1959-005G
1,6-Bis(diphenylphosphino)hexane ≥95.0%
1,6-Bis(diphenylphosphino)hexane

CAS Number: 19845-69-3
MDL Number: MFCD00003053
Molecular Formula: C30H32P2
Molecular Weight: 454.53
Purity/Analysis Method: >95.0% (GC,T)
Form: Crystal
Melting point (°C): 126


Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, Ligands
Other Product Names / Synonyms: Hexamethylenebis(diphenylphosphine)

Formula: C₃₀H₃₂P₂
MW: 454.53 g/mol
Melting Pt: 126 °C
MDL Number: MFCD00003053
CAS Number: 19845-69-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af320cdbfc5783fab1e12003d413fb070030cbc6&vid=af320cdbfc5783fab1e12003d413fb070030cbc6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR