4-Butyrylbiphenyl ≥99.0%

Supplier: TCI America

Synonyms: 4'-Phenylbutyrophenone

B1956-25G B1956-500G
TCB1956-025GEA 45.82 CAD
TCB1956-025G TCB1956-500G
4-Butyrylbiphenyl ≥99.0%
4-Butyrylbiphenyl

CAS Number: 13211-01-3
MDL Number: MFCD00022005
Molecular Formula: C16H16O
Molecular Weight: 224.30
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 355
Melting point (°C): 95


Other Product Names / Synonyms: 4'-Phenylbutyrophenone

Formula: C₁₆H₁₆O
MW: 224.30 g/mol
Boiling Pt: 355 °C
Melting Pt: 95 °C
MDL Number: MFCD00022005
CAS Number: 13211-01-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de6c571ed9be35bb7b81724675720cb91b221489&vid=de6c571ed9be35bb7b81724675720cb91b221489&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR