4-(2-Bromoethyl)benzoic acid methyl ester ≥98.0%

Supplier: TCI America

Synonyms: 4-(2-Bromoethyl)benzoic acid

B1944-25G B1944-5G
TCB1944-25GEA 1550.45 CAD
TCB1944-25G TCB1944-005G
4-(2-Bromoethyl)benzoic acid methyl ester ≥98.0%
4-(2-Bromoethyl)benzoic acid methyl ester

CAS Number: 52062-92-7
MDL Number: MFCD00016541
Molecular Formula: C9H9BrO2
Molecular Weight: 229.07
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 206

Formula: C₉H₉BrO₂
MW: 229.0706 g/mol
Melting Pt: 206 °C
MDL Number: MFCD00016541
CAS Number: 52062-92-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce79a05ef13861acf78337691a2b855ca3a00936&vid=ce79a05ef13861acf78337691a2b855ca3a00936&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR