(2-Benzothiazolylthio)acetic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B1941-25G B1941-500G
TCB1941-025GEA 42.66 CAD
TCB1941-025G TCB1941-500G
(2-Benzothiazolylthio)acetic acid ≥98.0% (by titrimetric analysis)
(2-Benzothiazolylthio)acetic acid

CAS Number: 6295-57-4
MDL Number: MFCD00227391
Molecular Formula: C9H7NO2S2
Molecular Weight: 225.28
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 159
Flash Point (°C): 235

Formula: C₉H₇NO₂S₂
Melting Pt: 159 °C
Flash Pt: 235 °C
MDL Number: MFCD00227391
CAS Number: 6295-57-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95d1118606fd70ee4d8ac2ef0e6619231604d707&vid=95d1118606fd70ee4d8ac2ef0e6619231604d707&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR