3-Benzylthiazolium bromide ≥98.0%

Supplier: TCI America

Synonyms: 3-Benzylthiazol-3-ium bromide

B1939-5G B1939-25G
TCB1939-005GEA 211.56 CAD
TCB1939-005G TCB1939-25G
3-Benzylthiazolium bromide ≥98.0%
3-Benzylthiazolium bromide

CAS Number: 75066-50-1
MDL Number: MFCD01631293
Molecular Formula: C10H10BrNS
Molecular Weight: 256.16
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 158

Formula: C₁₀H₁₀BrNS
MW: 256.16 g/mol
Melting Pt: 158 °C
MDL Number: MFCD01631293
CAS Number: 75066-50-1

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR