Benzyl chloromethyl sulfide ≥96.0%

Supplier: TCI America

Synonyms: alpha-(Chloromethylthio)toluene

B1937-5G B1937-25G
TCB1937-5GEA 67.78 CAD
TCB1937-5G TCB1937-025G
Benzyl chloromethyl sulfide ≥96.0%
Benzyl chloromethyl sulfide

CAS Number: 3970-13-6
MDL Number: MFCD02093436
Molecular Formula: C8H9ClS
Molecular Weight: 172.67
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.19
Storage Temperature: 0-10°C


Other Product Names / Synonyms: alpha-(Chloromethylthio)toluene

Formula: C₈H₉ClS
MW: 172.67 g/mol
Density: 1.19 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD02093436
CAS Number: 3970-13-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b1c1fbea8497206b1f7bd697f04d52e546adc95&vid=7b1c1fbea8497206b1f7bd697f04d52e546adc95&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR