2,6-Bis(bromomethyl)pyridine ≥99.0% (by GC, titration analysis)

Supplier: TCI America

B1936-5G B1936-1G
TCB1936-005GEA 361.82 CAD
TCB1936-005G TCB1936-001G
2,6-Bis(bromomethyl)pyridine ≥99.0% (by GC, titration analysis)
2,6-Bis(bromomethyl)pyridine

CAS Number: 7703-74-4
MDL Number: MFCD00191795
Molecular Formula: C7H7Br2N
Molecular Weight: 264.95
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 87

Formula: C₇H₇Br₂N
Melting Pt: 87 °C
MDL Number: MFCD00191795
CAS Number: 7703-74-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5897985c768b6003076cb3845c86cdee85d8d636&vid=5897985c768b6003076cb3845c86cdee85d8d636&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR