4-Butylcyclohexanol (cis- and trans- mixture) ≥98.0%

Supplier: TCI America

Synonyms: 4-Butylcyclohexanol

B1930-5G B1930-25G
TCB1930-5GEA 200.66 CAD
TCB1930-5G TCB1930-025G
4-Butylcyclohexanol (cis- and trans- mixture) ≥98.0%
4-Butylcyclohexanol (cis- and trans- mixture)

CAS Number: 70568-60-4
MDL Number: MFCD01321147
Molecular Formula: C10H20O
Molecular Weight: 156.27
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 122
Specific Gravity (20/20): 0.91

Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Boiling Pt: 122 °C
Density: 0.91 (20 °C)
MDL Number: MFCD01321147
CAS Number: 70568-60-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bfa3f0a59e9ea06aafdfec378211c554a3d2e277&vid=bfa3f0a59e9ea06aafdfec378211c554a3d2e277&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR