4-Octadecyloxybiphenyl ≥98.0%

Supplier: TCI America

B1924-25G B1924-5G
TCB1924-025GEA 198.29 CAD
TCB1924-025G TCB1924-5G
4-Octadecyloxybiphenyl ≥98.0%
4-Octadecyloxybiphenyl

CAS Number: 376609-78-8
MDL Number: MFCD01321145
Molecular Formula: C30H46O
Molecular Weight: 422.70
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 92


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: Biphenyl-4-yl Octadecyl Ether, Biphenyl-4-yl Stearyl Ether, 4-Stearyloxybiphenyl

Formula: C₃₀H₄₆O
Melting Pt: 92 °C
MDL Number: MFCD01321145
CAS Number: 376609-78-8

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR