4,4'-Bis(diethylphosphonomethyl)biphenyl ≥95.0% (by HPLC)
Supplier: TCI America
B1923-5G
B1923-1G
TCB1923-5GEA
64.46
CAD
TCB1923-5G
TCB1923-001G
4,4'-Bis(diethylphosphonomethyl)biphenyl ≥95.0% (by HPLC)
4,4'-Bis(diethylphosphonomethyl)biphenyl
CAS Number: 17919-34-5
MDL Number: MFCD01321144
Molecular Formula: C22H32O6P2
Molecular Weight: 454.44
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: [4,4'-Biphenylylenebis(methylene)]bisphosphonic Acid Tetraethyl Ester, Tetraethyl [4,4'-Biphenylylenebis(methylene)]bisphosphonate
Formula:
C₂₂H₃₂O₆P₂ |
MDL Number:
MFCD01321144 CAS Number: 17919-34-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...