N-Benzoyl-L-tyrosine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B1911-5G B1911-25G
TCB1911-005GEA 221.52 CAD
TCB1911-005G TCB1911-25G
N-Benzoyl-L-tyrosine ≥98.0% (by HPLC, titration analysis)
N-Benzoyl-L-tyrosine

CAS Number: 2566-23-6
MDL Number: MFCD01321140
Molecular Formula: C16H15NO4
Molecular Weight: 285.30
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: -77.5 deg (C=1, DMF)

Formula: C₁₆H₁₅NO₄
MDL Number: MFCD01321140
CAS Number: 2566-23-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa2bcee1221dc1c8378aaf801527c71ab32966b8&vid=aa2bcee1221dc1c8378aaf801527c71ab32966b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR