Benzeneruthenium(II) chloride dimer ≥95.0%

Supplier: TCI America

Synonyms: Dichloro(benzene)ruthenium(II) dimer

B1902-5G B1902-1G
TCB1902-5GEA 720.48 CAD
TCB1902-5G TCB1902-001G
Benzeneruthenium(II) chloride dimer ≥95.0%
Benzeneruthenium(II) chloride dimer

CAS Number: 37366-09-9
MDL Number: MFCD00064686
Molecular Formula: C12H12Cl4Ru2
Molecular Weight: 500.17
Purity/Analysis Method: >95.0% (T)
Form: Crystal

Formula: C₁₂H₁₂Cl₄Ru₂
MW: 500.17 g/mol
MDL Number: MFCD00064686
CAS Number: 37366-09-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d618c028fa89e74a8d7e4c3f3f92518fbf894b03&vid=d618c028fa89e74a8d7e4c3f3f92518fbf894b03&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR