3-Bromo-1-(triisopropylsilyl)pyrrole ≥95.0%
Supplier: TCI America
B1899-1G
B1899-25G
B1899-5G
TCB1899-001GEA
105.86
CAD
TCB1899-001G
TCB1899-25G
TCB1899-5G
3-Bromo-1-(triisopropylsilyl)pyrrole ≥95.0%
3-Bromo-1-(triisopropylsilyl)pyrrole
CAS Number: 87630-36-2
MDL Number: MFCD01321136
Molecular Formula: C13H24BrNSi
Molecular Weight: 302.33
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.16
Formula:
C₁₃H₂₄BrNSi Density: 1.16 (20 °C) |
MDL Number:
MFCD01321136 CAS Number: 87630-36-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...