5-Bromo-2,2'-bithiophene-5'-carboxaldehyde ≥98.0% (by HPLC)

Supplier: TCI America

B1874-5G B1874-1G
TCB1874-005GEA 358.66 CAD
TCB1874-005G TCB1874-001G
5-Bromo-2,2'-bithiophene-5'-carboxaldehyde ≥98.0% (by HPLC)
5-Bromo-2,2'-bithiophene-5'-carboxaldehyde

CAS Number: 110046-60-1
MDL Number: MFCD01321134
Molecular Formula: C9H5BrOS2
Molecular Weight: 273.16
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 147


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 5-Bromo-5'-formyl-2,2'-bithiophene

Formula: C₉H₅BrOS₂
Melting Pt: 147 °C
MDL Number: MFCD01321134
CAS Number: 110046-60-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=752bb407e80cf1c8f2671d605084522d019dcccb&vid=752bb407e80cf1c8f2671d605084522d019dcccb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR