1,3-Bis(tosyloxy)-2,2-dimethylpropane ≥98.0%
Supplier: TCI America
Synonyms:
1,3-Bis(p-toluenesulphonyloxy)-2,2-dimethylpropane, 1,3-Bis(p-toluenesulfonyloxy)-2,2-dimethylpropane, 2,2-Dimethyl-1,3-propanediol Ditosylate, Neopentyl Glycol Ditosylate
CAS Number: 22308-12-9
MDL Number: MFCD00110766
Molecular Formula: C19H24O6S2
Molecular Weight: 412.52
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 121
Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: 1,3-Bis(p-toluenesulfonyloxy)-2,2-dimethylpropane, 2,2-Dimethyl-1,3-propanediol Ditosylate, Neopentyl Glycol Ditosylate
Formula:
C₁₉H₂₄O₆S₂ MW: 412.52 g/mol Melting Pt: 121 °C |
MDL Number:
MFCD00110766 CAS Number: 22308-12-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...