1,2-Bis(3,4-dimethylphenyl)ethane ≥95.0%

Supplier: TCI America

B1851-1G B1851-5G
TCB1851-001GEA 200.03 CAD
TCB1851-001G TCB1851-005G
1,2-Bis(3,4-dimethylphenyl)ethane ≥95.0%
1,2-Bis(3,4-dimethylphenyl)ethane

CAS Number: 34101-86-5
MDL Number: MFCD00037055
Molecular Formula: C18H22
Molecular Weight: 238.37
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 94


Other Product Names / Synonyms: 1,2-Di(3,4-xylyl)ethane

Formula: C₁₈H₂₂
Melting Pt: 94 °C
MDL Number: MFCD00037055
CAS Number: 34101-86-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b46552fdb4e8211f9d31deb78d17529a22d9320&vid=9b46552fdb4e8211f9d31deb78d17529a22d9320&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR