4-Phenylbenzyl bromide ≥98.0%

Supplier: TCI America

Synonyms: 4-(Bromomethyl)biphenyl, 4-(Bromomethyl)-1,1'-biphenyl, p-(Bromomethyl)biphenyl

B1847-5G B1847-25G
TCB1847-005GEA 153.26 CAD
TCB1847-005G TCB1847-025G
4-Phenylbenzyl bromide ≥98.0%
4-Phenylbenzyl bromide

CAS Number: 2567-29-5
MDL Number: MFCD00017869
Molecular Formula: C13H11Br
Molecular Weight: 247.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 140
Melting point (°C): 85
Flash Point (°C): 110


Other Product Names / Synonyms: 4-Phenylbenzyl Bromide

Formula: C₁₃H₁₁Br
MW: 247.13 g/mol
Boiling Pt: 140 °C
Melting Pt: 85 °C
Flash Pt: 110 °C
MDL Number: MFCD00017869
CAS Number: 2567-29-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a24c01b09b2359c85ec89066751ca36e4ec961ae&vid=a24c01b09b2359c85ec89066751ca36e4ec961ae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR