Ethyl-2-(bromomethyl)acrylate ≥97.0% stabilized

Supplier: TCI America

B1820-1G B1820-5G
TCB1820-001GEA 88.43 CAD
TCB1820-001G TCB1820-005G
Ethyl-2-(bromomethyl)acrylate ≥97.0% stabilized

(stabilized with HQ)
CAS Number: 17435-72-2
MDL Number: MFCD00031518
Molecular Formula: C6H9BrO2
Molecular Weight: 193.04
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 86
Flash Point (°C): 81
Specific Gravity (20/20): 1.40
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-(Bromomethyl)acrylic Acid Ethyl Ester

Formula: C₆H₉BrO₂
Boiling Pt: 86 °C
Density: 1.40 (20 °C)
Flash Pt: 81 °C
MDL Number: MFCD00031518
CAS Number: 17435-72-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0c6ab3d119b1736a5ad52a53b07c982707c827a&vid=a0c6ab3d119b1736a5ad52a53b07c982707c827a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR