4-tert-Butylbiphenyl ≥98.0%

Supplier: TCI America

B1811-5G B1811-25G
TCB1811-5GEA 180.12 CAD
TCB1811-5G TCB1811-025G
4-tert-Butylbiphenyl ≥98.0%
4-tert-Butylbiphenyl

CAS Number: 1625-92-9
MDL Number: MFCD00222366
Molecular Formula: C16H18
Molecular Weight: 210.32
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 310
Melting point (°C): 52

Formula: C₁₆H₁₈
Boiling Pt: 310 °C
Melting Pt: 52 °C
MDL Number: MFCD00222366
CAS Number: 1625-92-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6e1354f2f22513537566d9708e15eebe2573d2c&vid=e6e1354f2f22513537566d9708e15eebe2573d2c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR