4-Bromomethyl-2-cyanobiphenyl ≥98.0%

Supplier: TCI America

Synonyms: 4'-(Bromomethyl)-[1,1'-biphenyl]-2-carbonitrile

B1806-25G B1806-5G
TCB1806-025GEA 173.8 CAD
TCB1806-025G TCB1806-005G
4-Bromomethyl-2-cyanobiphenyl ≥98.0%
4-Bromomethyl-2-cyanobiphenyl

CAS Number: 114772-54-2
MDL Number: MFCD00671503
Molecular Formula: C14H10BrN
Molecular Weight: 272.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 127


Other Product Names / Synonyms: 4'-Bromomethylbiphenyl-2-carbonitrile, 2-Cyano-4'-bromomethylbiphenyl

Formula: C₁₄H₁₀BrN
Melting Pt: 127 °C
MDL Number: MFCD00671503
CAS Number: 114772-54-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14919052c4fdd7acb8751fa0ddebcb4b0578d51d&vid=14919052c4fdd7acb8751fa0ddebcb4b0578d51d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR