1-Bromo-4-dodecylbenzene ≥95.0%

Supplier: TCI America

B1802-5G B1802-25G
TCB1802-005GEA 237 CAD
TCB1802-005G TCB1802-25G
1-Bromo-4-dodecylbenzene ≥95.0%
1-Bromo-4-dodecylbenzene

CAS Number: 126930-72-1
MDL Number: MFCD00191386
Molecular Formula: C18H29Br
Molecular Weight: 325.33
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 191
Freezing point (°C): 17
Specific Gravity (20/20): 1.07


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4-Dodecylbromobenzene

Formula: C₁₈H₂₉Br
MW: 325.33 g/mol
Boiling Pt: 191 °C
Density: 1.07 (20 °C)
MDL Number: MFCD00191386
CAS Number: 126930-72-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f3801464739877da6506858d9fbc71455a605bd&vid=1f3801464739877da6506858d9fbc71455a605bd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR