tert-Butyl-N-(Benzyloxy)carbamate ≥98.0% (by total nitrogen basis)
Supplier: TCI America
B1799-25G
B1799-5G
TCB1799-25GEA
401.64
CAD
TCB1799-25G
TCB1799-005G
tert-Butyl-N-(Benzyloxy)carbamate ≥98.0% (by total nitrogen basis)
tert-Butyl-N-(Benzyloxy)carbamate
CAS Number: 79722-21-7
MDL Number: MFCD00191873
Molecular Formula: C12H17NO3
Molecular Weight: 223.27
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 48
Other Product Names / Synonyms: N-(Benzyloxy)carbamic Acid tert-Butyl Ester
Formula:
C₁₂H₁₇NO₃ Melting Pt: 48 °C |
MDL Number:
MFCD00191873 CAS Number: 79722-21-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...