Bis(2-morpholinoethyl)ether ≥85.0%

Supplier: TCI America

Synonyms: 4,4'-(Oxybis(ethane-2,1-diyl))dimorpholine, 4,4'-(3-Oxapentane-1,5-diyl)bismorpholine

B1784-500G B1784-25G
TCB1784-500GEA 445.56 CAD
TCB1784-500G TCB1784-025G
Bis(2-morpholinoethyl)ether ≥85.0%
Bis(2-morpholinoethyl)ether

CAS Number: 6425-39-4
MDL Number: MFCD00072740
Molecular Formula: C12H24N2O3
Molecular Weight: 244.34
Purity/Analysis Method: >85.0% (GC)
Form: Clear Liquid
Boiling point (°C): 192
Melting point (°C): -28
Flash Point (°C): 146
Specific Gravity (20/20): 1.06


Product Categories: Materials Chemistry, Polymerization Catalysts
Other Product Names / Synonyms: 4,4'-(3-Oxapentane-1,5-diyl)bismorpholine

Formula: C₁₂H₂₄N₂O₃
MW: 244.34 g/mol
Boiling Pt: 192 °C
Melting Pt: –28 °C
Density: 1.06 (20 °C)
Flash Pt: 146 °C
MDL Number: MFCD00072740
CAS Number: 6425-39-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6956cdffc93df7448fdd3a182593904ff46cf1c6&vid=6956cdffc93df7448fdd3a182593904ff46cf1c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 85.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR