meso-Butane-1,2,3,4-tetracarboxylic dianhydride ≥98.0%

Supplier: TCI America

B1770-25G B1770-5G
TCB1770-025GEA 398.16 CAD
TCB1770-025G TCB1770-5G
meso-Butane-1,2,3,4-tetracarboxylic dianhydride ≥98.0%
meso-Butane-1,2,3,4-tetracarboxylic dianhydride

CAS Number: 17309-39-6
MDL Number: MFCD00671500
Molecular Formula: C8H6O6
Molecular Weight: 198.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₈H₆O₆
MW: 198.13 g/mol
MDL Number: MFCD00671500
CAS Number: 17309-39-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ae97141d07b0414f30a2b521d8943f477e0d3bb&vid=2ae97141d07b0414f30a2b521d8943f477e0d3bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR