1,2-Bis(trimethylsilyloxy)cyclobutene ≥94.0%

Supplier: TCI America

Synonyms: 1,2-Bis((trimethylsilyl)oxy)cyclobut-1-ene

B1768-5G B1768-1G
TCB1768-5GEA 272.44 CAD
TCB1768-5G TCB1768-001G
1,2-Bis(trimethylsilyloxy)cyclobutene ≥94.0%
1,2-Bis(trimethylsilyloxy)cyclobutene

CAS Number: 17082-61-0
MDL Number: MFCD00010798
Molecular Formula: C10H22O2Si2
Molecular Weight: 230.45
Purity/Analysis Method: >94.0% (GC)
Form: Clear Liquid
Boiling point (°C): 59
Flash Point (°C): 61
Specific Gravity (20/20): 0.90

Formula: C₁₀H₂₂O₂Si₂
MW: 230.45 g/mol
Boiling Pt: 59 °C
Density: 0.90 (20 °C)
Flash Pt: 61 °C
MDL Number: MFCD00010798
CAS Number: 17082-61-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=00900eeda590e121e2edb21b2a438f1479218cd7&vid=00900eeda590e121e2edb21b2a438f1479218cd7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR