(S)-(-)-3-tert-Butoxycarbonyl-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine ≥96.0%

Supplier: TCI America

Synonyms: (S)-3-tert-Butoxycarbonyl-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine, (S)-(-)-3-Boc-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine, Methyl (S)-(-)-3-tert-Butoxycarbonyl-2,2-dimethyl-4-oxazolidinecarboxylate

B1755-5G B1755-25G B1755-1G
TCB1755-5GEA 200.03 CAD
TCB1755-5G TCB1755-25G TCB1755-001G
(S)-(-)-3-tert-Butoxycarbonyl-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine ≥96.0%
(S)-(-)-3-tert-Butoxycarbonyl-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine

CAS Number: 108149-60-6
MDL Number: MFCD00192279
Molecular Formula: C12H21NO5
Molecular Weight: 259.30
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Flash Point (°C): 93


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(-)-3-Boc-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine, Methyl (S)-(-)-3-tert-Butoxycarbonyl-2,2-dimethyl-4-oxazolidinecarboxylate

Formula: C₁₂H₂₁NO₅
MW: 259.30 g/mol
Flash Pt: 93 °C
MDL Number: MFCD00192279
CAS Number: 108149-60-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9bd6b47d1f5b0364f9d01deb419b36d2221cf19e&vid=9bd6b47d1f5b0364f9d01deb419b36d2221cf19e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR