1,3-Bis(vinylsulfonyl)-2-propanol ≥98.0%

Supplier: TCI America

B1745-5G B1745-1G
TCB1745-5GEA 156.42 CAD
TCB1745-5G TCB1745-001G
1,3-Bis(vinylsulfonyl)-2-propanol ≥98.0%
1,3-Bis(vinylsulfonyl)-2-propanol

CAS Number: 67006-32-0
MDL Number: MFCD00671498
Molecular Formula: C7H12O5S2
Molecular Weight: 240.29
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 96

Formula: C₇H₁₂O₅S₂
MW: 240.29 g/mol
Melting Pt: 96 °C
MDL Number: MFCD00671498
CAS Number: 67006-32-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28d72e6e62d3301b90f95f9ce577350dffd97255&vid=28d72e6e62d3301b90f95f9ce577350dffd97255&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR