N-tert-Butyl-ɑ-(4-pyridyl-1-oxide)nitrone 98,0

Supplier: TCI America

Synonyms: POBN, N-tert-Butyl-ɑ-(4-pyridyl)nitrone N'-Oxide, α-(4-Pyridyl-1-Oxide)-N-t-butylnitrone, N-(4-Pyridylmethylene)-tert-butylamine N,N'-Dioxide, ɑ-(4-Pyridyl N-Oxide)-N-tert-butylnitrone

B1735-1G B1735-5G
TCB1735-001GEA 355.5 CAD
TCB1735-001G TCB1735-5G
N-tert-Butyl-ɑ-(4-pyridyl-1-oxide)nitrone 98,0
N-tert-Butyl-alpha-(4-pyridyl-1-oxide)nitrone

CAS Number: 66893-81-0
MDL Number: MFCD00041022
Molecular Formula: C10H14N2O2
Molecular Weight: 194.23
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 185
Storage Temperature: 0-10°C


Product Categories: Analytical Science, Spin Trapping Reagents
Other Product Names / Synonyms: N-tert-Butyl-alpha-(4-pyridyl)nitrone N'-Oxide, POBN, N-(4-Pyridylmethylene)-tert-butylamine N,N'-Dioxide, alpha-(4-Pyridyl N-Oxide)-N-tert-butylnitrone

Formula: C₁₀H₁₄N₂O₂
MW: 194.23 g/mol
Melting Pt: 185 °C
MDL Number: MFCD00041022
CAS Number: 66893-81-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b013068410e8656de6a6986b14b12d4f70c0dabd&vid=b013068410e8656de6a6986b14b12d4f70c0dabd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR