Bis(4-diethylaminophenyl)methanol ≥98.0% (by titrimetric analysis)
Supplier: TCI America
B1725-1G
B1725-5G
TCB1725-010GEA
563.08
CAD
TCB1725-010G
TCB1725-1G
TCB1725-5G
Bis(4-diethylaminophenyl)methanol ≥98.0% (by titrimetric analysis)
Bis(4-diethylaminophenyl)methanol
CAS Number: 134-91-8
MDL Number: MFCD00035979
Molecular Formula: C21H30N2O
Molecular Weight: 326.48
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 82
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 4,4'-Bis(diethylamino)benzhydrol
Formula:
C₂₁H₃₀N₂O Melting Pt: 82 °C |
MDL Number:
MFCD00035979 CAS Number: 134-91-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...